Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Zfand5
- Entrez Gene ID
-
22682
- Gene Name
- zinc finger, AN1-type domain 5
- Gene Aliases
-
2310057A04Rik, 5830475F03Rik, AA415485, Za20d2, Zfp216
- Chromosome Location
-
Chr.19: 21271279 - 21286840 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_009551.5 |
NP_033577.1 |
6 - 6 |
4007 |
75 |
GenBank mRNA |
AK009947.1 |
- |
1 - 1 |
1349 |
75 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CO430396.1 |
- |
1 |
488 |
|
Chr. 19 - 21282976 - 21283709 [+] on Build GRCm38 |
75 |
GenBankmRNA |
BM934826.1 |
- |
1 |
|
|
Chr. 19 - 21283107 - 21283764 [-] on Build GRCm38 |
75 |
GenBankmRNA |
CX729734.1 |
- |
1 |
373 |
|
Chr. 19 - 21283090 - 21283699 [+] on Build GRCm38 |
75 |
GenBankmRNA |
BP764354.1 |
- |
1 |
|
|
Chr. 19 - 21283229 - 21283769 [-] on Build GRCm38 |
75 |
GenBankmRNA |
CN458386.1 |
- |
1 |
562 |
|
Chr. 19 - 21282901 - 21283643 [+] on Build GRCm38 |
75 |
GenBankmRNA |
AW743675.1 |
- |
1 |
|
|
Chr. 19 - 21283325 - 21283766 [-] on Build GRCm38 |
75 |
GenBankmRNA |
AW744618.1 |
- |
1 |
|
|
Chr. 19 - 21283199 - 21283766 [-] on Build GRCm38 |
75 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC105242408 |
LOC105242408 |
|
-71.4kb
|
Anti-sense |
Chr. - 21165087 - 21210706 on Build GRCm38 |
Downstream |
guanine deaminase |
Gda |
AU015411, AW047581 |
107.5kb
|
Anti-sense |
Chr. - 21391307 - 21472661 on Build GRCm38 |
View Full Product Details