Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fem1c
Entrez Gene ID
240263
Gene Name
fem-1 homolog c (C.elegans)
Gene Aliases
2610312A07Rik, 3632443A22Rik, AI646567
Chromosome Location
Chr.18: 46504606 - 46525971 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK011987.1 - - 1552 76


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BE950795.1 - 1 Chr. 18 - 46501746 - 46502167 [+] on Build GRCm38 76
GenBankmRNA AI645205.1 - 1 314 Chr. 18 - 46501886 - 46502436 [-] on Build GRCm38 76
GenBankmRNA AL024126.1 - 1 382 Chr. 18 - 46501955 - 46502492 [-] on Build GRCm38 76
GenBankmRNA AV120230.2 - 1 615 Chr. 18 - 46501752 - 46502694 [-] on Build GRCm38 76
GenBankmRNA AW909474.1 - 1 Chr. 18 - 46501747 - 46502223 [+] on Build GRCm38 76
GenBankmRNA AW913080.1 - 1 147 Chr. 18 - 46501775 - 46502269 [-] on Build GRCm38 76
GenBankmRNA BB668741.1 - 1 43 Chr. 18 - 46501749 - 46502165 [-] on Build GRCm38 76
GenBankmRNA CO429258.1 - 1 482 Chr. 18 - 46501890 - 46502604 [-] on Build GRCm38 76
GenBankmRNA CN715002.1 - 1 576 Chr. 18 - 46502079 - 46502698 [-] on Build GRCm38 76
GenBankmRNA AI664085.1 - 1 216 Chr. 18 - 46501843 - 46502339 [-] on Build GRCm38 76
GenBankmRNA BB692475.1 - 1 57 Chr. 18 - 46501749 - 46502179 [-] on Build GRCm38 76
GenBankmRNA BX511913.1 - 1 148 Chr. 18 - 46501746 - 46502269 [-] on Build GRCm38 76


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream motile sperm domain containing 4 Mospd4 2010013H21Rik -35.9kb Sense Chr. - 46464934 - 46465816 on Build GRCm38

View Full Product Details