Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Phactr4
- Entrez Gene ID
-
100169
- Gene Name
- phosphatase and actin regulator 4
- Gene Aliases
-
3110001B12Rik, AI527228, AW495572, C330013F19Rik, N28169, mKIAA4120
- Chromosome Location
-
Chr.4: 132355922 - 132422446 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK013935.1 |
- |
1 - 1 |
1344 |
94 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
small nucleolar RNA host gene 3 |
Snhg3 |
Rnu17d, U17HG |
-33.3kb
|
Sense |
Chr. - 132351933 - 132353686 on Build GRCm38 |
Downstream |
mediator complex subunit 18 |
Med18 |
2810046C01Rik |
70.2kb
|
Sense |
Chr. - 132458729 - 132463921 on Build GRCm38 |
View Full Product Details