Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Rian
- Entrez Gene ID
-
75745
- Gene Name
- RNA imprinted and accumulated in nucleus
- Gene Aliases
-
5530401N18Rik, C130089L09Rik, Meg8
- Chromosome Location
-
Chr.12: 109603945 - 109661716 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_028261.1 |
|
10 - 11 |
927 |
105 |
GenBank mRNA |
AF498301.1 |
- |
7 - 8 |
524 |
105 |
AF498302.1 |
- |
7 - 8 |
573 |
105 |
EU434919.1 |
- |
14 - 15 |
1246 |
105 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
snoRNA AF357426 |
AF357426 |
MBII-49 |
-2.4kb
|
Sense |
Chr. - 109642838 - 109642904 on Build GRCm38 |
Downstream |
snoRNA DQ267100 |
DQ267100 |
|
3.3kb
|
Sense |
Chr. - 109649617 - 109649687 on Build GRCm38 |
View Full Product Details