Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Actr1a
- Entrez Gene ID
-
54130
- Gene Name
- ARP1 actin-related protein 1A, centractin alpha
- Gene Aliases
-
Arp1, alpha-Arp1
- Chromosome Location
-
Chr.19: 46376814 - 46395735 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK018927.1 |
- |
1 - 1 |
447 |
67 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BX640028.1 |
- |
1 |
|
|
Chr. 19 - 46395655 - 46395954 [-] on Build GRCm38 |
67 |
GenBankmRNA |
BU582895.1 |
- |
1 |
450 |
|
Chr. 19 - 46395373 - 46395926 [+] on Build GRCm38 |
67 |
GenBankmRNA |
CF105069.1 |
- |
1 |
444 |
|
Chr. 19 - 46395379 - 46395947 [+] on Build GRCm38 |
67 |
GenBankmRNA |
BF018834.1 |
- |
1 |
|
|
Chr. 19 - 46395500 - 46395954 [-] on Build GRCm38 |
67 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
transmembrane protein 180 |
Tmem180 |
4930449A08Rik, 4930538D17Rik |
-20.1kb
|
Sense |
Chr. - 46356877 - 46375254 on Build GRCm38 |
Downstream |
suppressor of fused homolog (Drosophila) |
Sufu |
Su(fu) |
949.0
|
Sense |
Chr. - 46396896 - 46488804 on Build GRCm38 |
View Full Product Details