Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Atxn1
- Entrez Gene ID
-
20238
- Gene Name
- ataxin 1
- Gene Aliases
-
2900016G23Rik, Atx1, C85907, ENSMUSG00000074917, Gm10786, Sca1
- Chromosome Location
-
Chr.13: 45549755 - 45965036 on Build GRCm38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BY712500.1 |
- |
1 |
346 |
|
Chr. 13 - 45552673 - 45553124 [-] on Build GRCm38 |
71 |
GenBankmRNA |
CB237212.1 |
- |
|
242 |
|
Chr. - - [-] on Build |
71 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
guanosine monophosphate reductase |
Gmpr |
2310004P21Rik, AV028449 |
-6.3kb
|
Anti-sense |
Chr. - 45507429 - 45546386 on Build GRCm38 |
Downstream |
RIKEN cDNA 5033430I15 gene |
5033430I15Rik |
|
411.9kb
|
Anti-sense |
Chr. - 45965060 - 45966508 on Build GRCm38 |
View Full Product Details