Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Katna1
- Entrez Gene ID
-
23924
- Gene Name
- katanin p60 (ATPase-containing) subunit A1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.10: 7726000 - 7763150 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK132823.1 |
- |
1 - 2 |
164 |
83 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
large tumor suppressor |
Lats1 |
AW208599 |
-27.1kb
|
Anti-sense |
Chr. - 7681209 - 7716461 on Build GRCm38 |
Downstream |
glycoprotein integral membrane 1 |
Ginm1 |
AA589616 |
6.8kb
|
Sense |
Chr. - 7767947 - 7780946 on Build GRCm38 |
View Full Product Details