Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Gm34888
- Entrez Gene ID
-
102638288
- Gene Name
- predicted gene, 34888
- Gene Aliases
-
-
- Chromosome Location
-
Chr.9: 109536660 - 109554346 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK034386.1 |
- |
2 - 3 |
272 |
69 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BB626421.1 |
- |
2 - 3 |
272 |
|
Chr. 9 - 109537100 - 109550818 [+] on Build GRCm38 |
69 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
F-box and WD-40 domain protein 23 |
Fbxw23 |
Gm7163 |
-3.4kb
|
Anti-sense |
Chr. - 109516398 - 109535928 on Build GRCm38 |
Downstream |
F-box and WD-40 domain protein 15 |
Fbxw15 |
Fbxo12J |
7.4kb
|
Anti-sense |
Chr. - 109552602 - 109569438 on Build GRCm38 |
View Full Product Details