Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Otud3
Entrez Gene ID
73162
Gene Name
OTU domain containing 3
Gene Aliases
3110030K17Rik
Chromosome Location
Chr.4: 138891063 - 138913947 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK076876.1 - 1 - 1 753 73


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AI036198.1 - 1 78 Chr. 4 - 138891071 - 138891447 [-] on Build GRCm38 73
GenBankmRNA AV267369.2 - 1 347 Chr. 4 - 138891072 - 138891721 [-] on Build GRCm38 73
GenBankmRNA BB706496.1 - 1 37 Chr. 4 - 138891070 - 138891415 [-] on Build GRCm38 73
GenBankmRNA BE136210.1 - 1 Chr. 4 - 138891065 - 138891538 [+] on Build GRCm38 73
GenBankmRNA BM876712.1 - 1 Chr. 4 - 138891063 - 138891546 [+] on Build GRCm38 73
GenBankmRNA BB810207.1 - 1 102 Chr. 4 - 138891066 - 138891480 [-] on Build GRCm38 73
GenBankmRNA BX630174.1 - 1 Chr. 4 - 138891063 - 138891447 [+] on Build GRCm38 73


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream phospholipase A2, group IIE Pla2g2e -8.2kb Anti-sense Chr. - 138877911 - 138882817 on Build GRCm38
Downstream ring finger protein 186 Rnf186 9130020G10Rik 75.0kb Anti-sense Chr. - 138967112 - 138968366 on Build GRCm38

View Full Product Details