Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Sgcg
- Entrez Gene ID
-
24053
- Gene Name
- sarcoglycan, gamma (dystrophin-associated glycoprotein)
- Gene Aliases
-
35kDa, 5430420E18Rik, AI642964, gamma-SG
- Chromosome Location
-
Chr.14: 61219106 - 61274858 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK213459.1 |
- |
1 - 2 |
36 |
91 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
sacsin |
Sacs |
A230052M14, DNAJC29, E130115J16Rik, mKIAA0730 |
-24.3kb
|
Anti-sense |
Chr. - 61138427 - 61240693 on Build GRCm38 |
Downstream |
RIKEN cDNA 1700109G14 gene |
1700109G14Rik |
1700126G13Rik |
18.2kb
|
Sense |
Chr. - 61292983 - 61305334 on Build GRCm38 |
View Full Product Details