Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Triml2
Entrez Gene ID
622117
Gene Name
tripartite motif family-like 2
Gene Aliases
EG622117, Tu4
Chromosome Location
Chr.8: 43180506 - 43193884 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160412.1 NP_001153884.1 2 - 3 341 92

View Full Product Details