Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tgm7
Entrez Gene ID
640543
Gene Name
transglutaminase 7
Gene Aliases
TGMZ, TGz
Chromosome Location
Chr.2: 121093498 - 121127516 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160424.1 NP_001153896.1 6 - 7 1000 84

View Full Product Details