Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cers3
Entrez Gene ID
545975
Gene Name
ceramide synthase 3
Gene Aliases
4930550L11Rik, Gm488, Lass3, T3L
Chromosome Location
Chr.7: 66733178 - 66823697 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164201.1 NP_001157673.1 2 - 3 418 122
GenBank mRNA DQ358087.1 - 1 - 2 176 122
DQ646881.1 - 2 - 3 297 122

View Full Product Details