Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hephl1
Entrez Gene ID
244698
Gene Name
hephaestin-like 1
Gene Aliases
Gm509, Zp, zyklopen
Chromosome Location
Chr.9: 15051841 - 15112108 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164797.1 NP_001158269.1 14 - 15 2574 84
GenBank mRNA AK132457.1 - 14 - 15 2575 84
BB618242.1 - 1 - 2 92 84

View Full Product Details