The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001165941.1 | NP_001159413.1 | 2 - 3 | 509 | 94 |
NM_001165942.1 | NP_001159414.1 | 1 - 2 | 396 | 94 | |
NM_028950.4 | NP_083226.3 | 1 - 2 | 396 | 94 | |
GenBank mRNA | AK016811.1 | - | 2 - 3 | 522 | 94 |
AK042750.1 | - | 1 - 2 | 396 | 94 | |
AK077778.1 | - | 1 - 2 | 280 | 94 | |
AK132428.1 | - | 1 - 2 | 363 | 94 | |
BB639947.1 | - | 1 - 2 | 396 | 94 | |
BC053751.1 | - | 1 - 2 | 364 | 94 |