The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243307.1 | NP_001230236.1 | 2 - 2 | 507 | 100 |
NM_198616.4 | NP_941018.1 | 1 - 1 | 662 | 100 | |
GenBank mRNA | AK140354.1 | - | 1 - 1 | 996 | 100 |
BC058411.1 | - | 1 - 1 | 648 | 100 | |
BC094592.1 | - | 2 - 2 | 448 | 100 | |
BQ445696.1 | - | 1 - 1 | 648 | 100 |