Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hmga2
Entrez Gene ID
15364
Gene Name
high mobility group AT-hook 2
Gene Aliases
9430083A20Rik, HMGI-C, Hmgic, pg, pygmy
Chromosome Location
Chr.10: 120361275 - 120476935 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010441.2 NP_034571.1 1 - 1 449 110

View Full Product Details