The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029751.4 | NP_084027.1 | 4 - 5 | 502 | 114 |
GenBank mRNA | AK010983.1 | - | 4 - 5 | 473 | 114 |
AK146543.1 | - | 4 - 5 | 472 | 114 | |
AK150820.1 | - | 4 - 5 | 471 | 114 | |
AK152611.1 | - | 4 - 5 | 472 | 114 | |
AK161942.1 | - | 4 - 5 | 472 | 114 | |
AK162047.1 | - | 4 - 5 | 473 | 114 | |
AK168258.1 | - | 4 - 5 | 472 | 114 | |
BC024609.1 | - | 4 - 5 | 446 | 114 | |
BC037146.1 | - | 4 - 5 | 514 | 114 |