Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Amica1
Entrez Gene ID
270152
Gene Name
adhesion molecule, interacts with CXADR antigen 1
Gene Aliases
AMICA, Crea7, Gm638
Chromosome Location
Chr.9: 45079162 - 45108531 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005421.4 NP_001005421.3 1 - 2 113 88
GenBank mRNA AY093688.1 - 1 - 2 113 88
CJ139627.1 - 1 - 2 67 88

View Full Product Details