Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zbtb9
Entrez Gene ID
474156
Gene Name
zinc finger and BTB domain containing 9
Gene Aliases
3930402F13Rik
Chromosome Location
Chr.17: 26973179 - 26976203 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005916.2 NP_001005916.1 1 - 2 134 103
GenBank mRNA AK154850.1 - 1 - 2 116 103
BC049933.1 - 1 - 2 134 103

View Full Product Details