The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019405.6 | NP_062278.2 | 3 - 4 | 317 | 98 |
GenBank mRNA | AK004081.1 | - | 3 - 4 | 318 | 98 |
AK010541.1 | - | 3 - 4 | 308 | 98 | |
AK088488.1 | - | 1 - 2 | 149 | 98 | |
AK140156.1 | - | 3 - 4 | 335 | 98 | |
AK151033.1 | - | 3 - 4 | 385 | 98 | |
BC013545.1 | - | 2 - 3 | 301 | 98 |