The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019420.2 | NP_062293.1 | 1 - 1 | 1247 | 94 |
GenBank mRNA | AF082504.1 | - | 1 - 1 | 1240 | 94 |
AK007600.1 | - | 1 - 1 | 1235 | 94 | |
AK166333.1 | - | 1 - 1 | 1204 | 94 | |
KJ650213.1 | - | 1 - 1 | 1021 | 94 |