Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Plag1
Entrez Gene ID
56711
Gene Name
pleiomorphic adenoma gene 1
Gene Aliases
-
Chromosome Location
Chr.4: 3898769 - 3938434 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019969.3 NP_064353.2 1 - 2 192 73
GenBank mRNA AK044405.1 - 1 - 2 181 73
AY574219.1 - 1 - 2 169 73
BC139283.1 - 1 - 2 142 73
BC139285.1 - 1 - 2 142 73
CJ178210.1 - 1 - 2 193 73

View Full Product Details