Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccl26
Entrez Gene ID
541307
Gene Name
chemokine (C-C motif) ligand 26
Gene Aliases
Ccl26l
Chromosome Location
Chr.5: 135560448 - 135563569 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013412.2 NP_001013430.2 2 - 3 191 98

View Full Product Details