The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033332.2 | NP_001028504.1 | 3 - 4 | 630 | 84 |
NM_001110013.1 | NP_001103483.1 | 3 - 4 | 630 | 84 | |
GenBank mRNA | AK044857.1 | - | 3 - 4 | 586 | 84 |
AK149759.1 | - | 3 - 4 | 589 | 84 | |
AK158192.1 | - | 3 - 4 | 616 | 84 | |
AK165838.1 | - | 3 - 4 | 631 | 84 |