Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pkd1l3
Entrez Gene ID
244646
Gene Name
polycystic kidney disease 1 like 3
Gene Aliases
-
Chromosome Location
Chr.8: 109613994 - 109673526 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039700.2 NP_001034789.2 2 - 3 430 101
NM_001286454.1 NP_001273383.1 2 - 3 430 101
NM_181544.2 NP_853522.2 2 - 3 430 101
GenBank mRNA AB290926.1 - 2 - 3 430 101
AK044116.1 - 2 - 3 246 101
AY164486.1 - 2 - 3 430 101
DQ382344.1 - 2 - 3 430 101
DQ382345.1 - 2 - 3 430 101
DQ382346.1 - 3 - 4 442 101

View Full Product Details