The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039889.3 | NP_001034978.1 | 1 - 2 | 431 | 66 |
GenBank mRNA | AK133180.1 | - | 1 - 2 | 432 | 66 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001126045.1 | NP_001119517.1 | 1 - 2 | 257 | 66 |
NM_001168602.1 | NP_001162073.1 | 1 - 2 | 410 | 66 | |
GenBank mRNA | AJ245454.1 | - | 1 - 2 | 410 | 66 |
BC150960.1 | - | - | 257 | 66 |