Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Xlr5a
- Entrez Gene ID
-
574438
- Gene Name
- X-linked lymphocyte-regulated 5A
- Gene Aliases
-
Xlr5d
- Chromosome Location
-
Chr.X: 73107621 - 73119524 on Build GRCm38
- Gene Symbol
- Xlr5b
- Entrez Gene ID
-
627081
- Gene Name
- X-linked lymphocyte-regulated 5B
- Gene Aliases
-
OTTMUSG00000017646
- Chromosome Location
-
Chr.X: 73146976 - 73158886 on Build GRCm38
- Gene Symbol
- Xlr5c
- Entrez Gene ID
-
27084
- Gene Name
- X-linked lymphocyte-regulated 5C
- Gene Aliases
-
Xlr5
- Chromosome Location
-
Chr.X: 73283361 - 73293970 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_031493.1 |
NP_113681.1 |
5 - 6 |
436 |
95 |
GenBank mRNA |
BC119509.2 |
- |
5 - 6 |
436 |
95 |
View Full Product Details