The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001104557.1 | NP_001098027.1 | 1 - 2 | 63 | 85 |
GenBank mRNA | CK788351.1 | - | - | 53 | 85 |