Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vmn2r74
Entrez Gene ID
546980
Gene Name
vomeronasal 2, receptor 74
Gene Aliases
EG546980
Chromosome Location
Chr.7: 85951867 - 85961482 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105187.1 NP_001098657.1 2 - 3 479 81

View Full Product Details