The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009967.2 | NP_034097.1 | 2 - 3 | 320 | 105 |
GenBank mRNA | AF032995.1 | - | 2 - 3 | 316 | 105 |
AK143060.1 | - | 2 - 3 | 317 | 105 | |
BC056452.1 | - | 2 - 3 | 296 | 105 |