Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
H2-Q6
Entrez Gene ID
110557
Gene Name
histocompatibility 2, Q region locus 6
Gene Aliases
0610037M15Rik, H-2Q6, Qa-6, Qa6
Chromosome Location
Chr.17: 35424799 - 35430053 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207648.1 NP_997531.1 4 - 5 882 75

Gene Symbol
H2-Q8
Entrez Gene ID
15019
Gene Name
histocompatibility 2, Q region locus 8
Gene Aliases
H-2Q8, MMS10-T, Ms10t, Qa-2, Qa-8, Qa8
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023124.5 NP_075613.3 4 - 5 910 75

Gene Symbol
LOC68395
Entrez Gene ID
68395
Gene Name
histocompatibility 2, Q region locus 6-like
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC025170.1 - - 664 75

View Full Product Details