The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_207652.2 | NP_997535.2 | 2 - 3 | 3237 | 97 |
GenBank mRNA | AF201285.1 | - | 2 - 3 | 2948 | 97 |
AK173322.1 | - | 4 - 5 | 3649 | 97 | |
BC058660.1 | - | 2 - 3 | 3237 | 97 |