Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Gm20854
- Entrez Gene ID
-
100040911
- Gene Name
- predicted gene, 20854
- Gene Aliases
-
-
- Chromosome Location
-
Chr.Y: 85528523 - 85529519 on Build GRCm38
- Gene Symbol
- Gm20806
- Entrez Gene ID
-
100039574
- Gene Name
- predicted gene, 20806
- Gene Aliases
-
-
- Chromosome Location
-
Chr.Y: 78835721 - 78836719 on Build GRCm38
- Gene Symbol
- Gm20852
- Entrez Gene ID
-
100040786
- Gene Name
- predicted gene, 20852
- Gene Aliases
-
-
- Chromosome Location
-
Chr.Y: 66739797 - 66742170 on Build GRCm38
- Gene Symbol
- Gm20868
- Entrez Gene ID
-
100041284
- Gene Name
- predicted gene, 20868
- Gene Aliases
-
-
- Chromosome Location
-
Chr.Y: 71297832 - 71300148 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK006494.1 |
- |
2 - 2 |
969 |
74 |
- Gene Symbol
- Gm20917
- Entrez Gene ID
-
100042428
- Gene Name
- predicted gene, 20917
- Gene Aliases
-
-
- Chromosome Location
-
Chr.Y: 79148793 - 79149787 on Build GRCm38
- Gene Symbol
- Gm21394
- Entrez Gene ID
-
100040744
- Gene Name
- predicted gene, 21394
- Gene Aliases
-
-
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC100414.1 |
- |
2 - 2 |
979 |
74 |
- Gene Symbol
- Ssty2
- Entrez Gene ID
-
70009
- Gene Name
- spermiogenesis specific transcript on the Y 2
- Gene Aliases
-
1700029H17Rik, pc11
- Chromosome Location
-
Chr.Y: 67045743 - 67048072 on Build GRCm38
View Full Product Details