The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199015.4 | NP_950180.3 | 7 - 8 | 669 | 117 |
GenBank mRNA | AK156583.1 | - | 7 - 8 | 650 | 117 |
BC145673.1 | - | 7 - 8 | 589 | 117 | |
EU155107.1 | - | 4 - 5 | 280 | 117 |