The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166001.1 | NP_001159473.1 | 1 - 2 | 193 | 88 |
GenBank mRNA | BC137866.1 | - | 1 - 2 | 134 | 88 |
BC144985.1 | - | 1 - 2 | 134 | 88 |