The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166503.1 | NP_001159975.1 | 9 - 10 | 1102 | 84 |
NM_027216.5 | NP_081492.4 | 9 - 10 | 1081 | 84 | |
GenBank mRNA | AK007986.1 | - | 9 - 10 | 1021 | 84 |
AK036999.1 | - | 9 - 10 | 1028 | 84 | |
AK049368.1 | - | 9 - 10 | 1046 | 84 | |
AK165545.1 | - | 7 - 8 | 933 | 84 | |
AK170673.1 | - | 9 - 10 | 1021 | 84 | |
BC004643.1 | - | 5 - 6 | 611 | 84 | |
BC019647.1 | - | 9 - 10 | 1025 | 84 | |
BC031751.1 | - | 10 - 11 | 1139 | 84 |