The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007487.3 | NP_031513.1 | 1 - 2 | 116 | 90 |
GenBank mRNA | AK048606.1 | - | 1 - 2 | 117 | 90 |
AK137751.1 | - | - | 117 | 90 | |
AK168618.1 | - | 1 - 2 | 117 | 90 | |
BC003725.1 | - | - | 105 | 90 | |
BC029234.1 | - | 1 - 2 | 76 | 90 |