Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cela2a
Entrez Gene ID
13706
Gene Name
chymotrypsin-like elastase family, member 2A
Gene Aliases
Ela-2, Ela2, Ela2a
Chromosome Location
Chr.4: 141814963 - 141826003 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007919.2 NP_031945.1 4 - 5 379 83
GenBank mRNA BC026552.1 - 4 - 5 368 83
X04573.1 - 4 - 5 379 83

View Full Product Details