Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Foxl1
Entrez Gene ID
14241
Gene Name
forkhead box L1
Gene Aliases
FREAC7, Fkh6, fkh-6
Chromosome Location
Chr.8: 121127685 - 121130644 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008024.2 NP_032050.2 1 - 1 148 119

View Full Product Details