The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008631.2 | NP_032657.1 | 1 - 2 | 70 | 89 |
GenBank mRNA | AK131894.1 | - | 1 - 2 | 70 | 89 |