The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001277095.1 | NP_001264024.1 | 1 - 2 | 207 | 90 |
NM_008692.4 | NP_032718.2 | 1 - 2 | 207 | 90 | |
GenBank mRNA | AK011327.1 | - | 1 - 2 | 164 | 90 |
AK156353.1 | - | 1 - 2 | 61 | 90 | |
BC020117.1 | - | 1 - 2 | 133 | 90 | |
BC085261.1 | - | 1 - 2 | 133 | 90 | |
CJ092564.1 | - | 1 - 2 | 184 | 90 |