The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291508.1 | NP_001278437.1 | 1 - 2 | 858 | 81 |
NM_001291509.1 | NP_001278438.1 | 1 - 2 | 858 | 81 | |
NM_008783.3 | NP_032809.1 | 1 - 2 | 858 | 81 | |
NM_183355.3 | NP_899198.1 | 1 - 2 | 858 | 81 | |
GenBank mRNA | AB221563.1 | - | 1 - 2 | 193 | 81 |
AF020196.1 | - | 1 - 2 | 549 | 81 | |
AK037006.1 | - | 1 - 2 | 822 | 81 | |
AK045252.1 | - | 1 - 2 | 857 | 81 | |
AK084143.1 | - | 1 - 2 | 350 | 81 | |
AK144346.1 | - | 1 - 2 | 860 | 81 | |
AK144446.1 | - | 1 - 2 | 860 | 81 | |
BC002244.1 | - | 1 - 2 | 84 | 81 | |
BC054376.1 | - | 1 - 2 | 819 | 81 | |
BC058390.1 | - | 1 - 2 | 586 | 81 | |
HM369110.1 | - | 1 - 2 | 193 | 81 | |
L27453.1 | - | 1 - 2 | 237 | 81 |