Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rasa3
Entrez Gene ID
19414
Gene Name
RAS p21 protein activator 3
Gene Aliases
AI326412, C86362, E130011G04, GAPIII, scat
Chromosome Location
Chr.8: 13567218 - 13677587 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009025.2 NP_033051.2 12 - 13 1346 103
GenBank mRNA AK053353.1 - 12 - 13 1342 103
AK141511.1 - 12 - 13 1353 103
AK147896.1 - 12 - 13 1314 103
AK166354.1 - 12 - 13 1344 103
BC057300.1 - 12 - 13 1337 103
BC060195.1 - 12 - 13 1361 103
BC068297.1 - 12 - 13 1210 103
U20238.1 - 12 - 13 1331 103

View Full Product Details