Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Serpina1c
- Entrez Gene ID
-
20702
- Gene Name
- serine (or cysteine) peptidase inhibitor, clade A, member 1C
- Gene Aliases
-
Pi3, Pi6, Skalp, Spi1-3, Spi1-6, Wap3
- Chromosome Location
-
Chr.12: 103894926 - 103904950 on Build GRCm38
- Gene Symbol
- Serpina1a
- Entrez Gene ID
-
20700
- Gene Name
- serine (or cysteine) peptidase inhibitor, clade A, member 1A
- Gene Aliases
-
Aat-2, Aat2, Dom1, PI1, Spi1-1, Spi1-3
- Chromosome Location
-
Chr.12: 103853295 - 103863619 on Build GRCm38
View Full Product Details