Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccr2
Entrez Gene ID
12772
Gene Name
chemokine (C-C motif) receptor 2
Gene Aliases
Cc-ckr-2, Ccr2a, Ccr2b, Ckr2, Ckr2a, Ckr2b, Cmkbr2, mJe-r
Chromosome Location
Chr.9: 124101918 - 124109140 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009915.2 NP_034045.1 2 - 3 124 99
GenBank mRNA AK046579.1 - - 358 99
AK138599.1 - 1 - 2 437 99
AK154590.1 - 1 - 2 1873 99
BC138803.1 - 1 - 2 84 99
BC138804.1 - 1 - 2 84 99
U51717.1 - 1 - 2 82 99
U56819.1 - 2 - 3 124 99

View Full Product Details