Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Csn2
Entrez Gene ID
12991
Gene Name
casein beta
Gene Aliases
Csnb
Chromosome Location
Chr.5: 87692619 - 87699429 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009972.2 NP_034102.1 3 - 4 138 109
GenBank mRNA AK052803.1 - 3 - 4 135 109
AK052805.1 - 3 - 4 135 109
AK085694.1 - 3 - 4 135 109
AK085729.1 - 3 - 4 135 109
AK142616.1 - 3 - 4 135 109
AK142620.1 - 3 - 4 135 109
AK142631.1 - 3 - 4 135 109
AK142635.1 - 3 - 4 135 109
AK164747.1 - 3 - 4 135 109
AK164761.1 - 3 - 4 135 109
AK164791.1 - 3 - 4 135 109
AK164792.1 - 3 - 4 132 109
AK164793.1 - 3 - 4 132 109
AK164794.1 - 3 - 4 135 109
BC013332.1 - 3 - 4 135 109
BC020356.1 - 3 - 4 115 109
BC021153.1 - 3 - 4 133 109
BC050270.1 - 3 - 4 115 109
BC092098.1 - 3 - 4 112 109
X04490.1 - 3 - 4 134 109

View Full Product Details