Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Pira1
- Entrez Gene ID
-
18722
- Gene Name
- paired-Ig-like receptor A1
- Gene Aliases
-
6M21, Ly89, PIR-A1, Pir
- Chromosome Location
-
Chr.7: 3731633 - 3739870 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_011087.1 |
NP_035217.1 |
- |
1917 |
82 |
GenBank mRNA |
U96682.1 |
- |
- |
1917 |
82 |
- Gene Symbol
- Pira11
- Entrez Gene ID
-
18724
- Gene Name
- paired-Ig-like receptor A11
- Gene Aliases
-
Gm14548, p91C
- Chromosome Location
-
-
- Gene Symbol
- Pira2
- Entrez Gene ID
-
18725
- Gene Name
- paired-Ig-like receptor A2
- Gene Aliases
-
6M23
- Chromosome Location
-
Chr.7: 3836810 - 3845099 on Build GRCm38
- Gene Symbol
- Pira4
- Entrez Gene ID
-
18727
- Gene Name
- paired-Ig-like receptor A4
- Gene Aliases
-
6M7
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_011091.1 |
NP_035221.1 |
- |
1916 |
82 |
GenBank mRNA |
U96685.1 |
- |
- |
1916 |
82 |
- Gene Symbol
- Pira5
- Entrez Gene ID
-
18728
- Gene Name
- paired-Ig-like receptor A5
- Gene Aliases
-
7M4
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
U96686.1 |
- |
9 - 10 |
1620 |
82 |
- Gene Symbol
- Pira7
- Entrez Gene ID
-
18730
- Gene Name
- paired-Ig-like receptor A7
- Gene Aliases
-
16M2
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_011094.1 |
NP_035224.1 |
10 - 11 |
1944 |
82 |
GenBank mRNA |
U96688.1 |
- |
10 - 11 |
1944 |
82 |
- Gene Symbol
- Pirb
- Entrez Gene ID
-
18733
- Gene Name
- paired Ig-like receptor B
- Gene Aliases
-
Gp91, LIR-3, Lilrb3, PIR-B
- Chromosome Location
-
Chr.7: 3712505 - 3720456 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK165650.1 |
- |
- |
2093 |
82 |
View Full Product Details