Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mxi1
Entrez Gene ID
17859
Gene Name
Max interacting protein 1
Gene Aliases
ENSMUSG00000067085, Gm10197, Mad2, bHLHc11
Chromosome Location
Chr.19: 53310371 - 53375810 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010847.3 NP_034977.1 1 - 2 309 62
GenBank mRNA AK017637.1 - 1 - 2 262 62
AK140465.1 - 1 - 2 259 62
L38822.1 - 1 - 2 181 62

View Full Product Details