The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025407.2 | NP_079683.2 | 1 - 2 | 142 | 71 |
GenBank mRNA | AK004031.1 | - | 1 - 2 | 89 | 71 |
AK010553.1 | - | 1 - 2 | 82 | 71 | |
AK013128.1 | - | 1 - 2 | 82 | 71 | |
AK151764.1 | - | 1 - 2 | 84 | 71 | |
AK160337.1 | - | 1 - 2 | 89 | 71 | |
AK167911.1 | - | 1 - 2 | 82 | 71 | |
AK168218.1 | - | 1 - 2 | 84 | 71 | |
BB649589.1 | - | 1 - 2 | 142 | 71 | |
BC030064.1 | - | 1 - 2 | 79 | 71 | |
BY260300.1 | - | 1 - 2 | 115 | 71 |